missing translation for 'onlineSavingsMsg'
Learn More

CLOCK Antibody [PerCP], Novus Biologicals™

Sheep Polyclonal Antibody

Brand:  Novus Biologicals AF4026C

Product Code. 30658423

  • 638.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CLOCK Polyclonal antibody specifically detects CLOCK in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLOCK
Polyclonal
PBS
Sheep
Antigen Affinity-purified
RUO
Primary
Human
Purified
Western Blot
PerCP
BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein
E. coli-derived recombinant human CLOCK, Glu108-Glu234, Accession # O15516
0.1 mL
Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
9575
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.