missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Antibody [FITC], Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals AF4026F
This item is not returnable.
View return policy
Description
CLOCK Polyclonal antibody specifically detects CLOCK in Human samples. It is validated for Western Blot
Specifications
| CLOCK | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| FITC | |
| BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
| E. coli-derived recombinant human CLOCK, Glu108-Glu234, Accession # O15516 | |
| 0.1 mL | |
| Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision | |
| 9575 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction