Learn More
Abnova™ CLNS1A (Human) Recombinant Protein
Human CLNS1A full-length ORF ( NP_001284, 1 a.a. - 237 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001207-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSFLKSFPPPGPAEGLLRQQPDTEAVLNGKGLGTGTLYIAESRLSWLDGSGLGFSLEYPTISLHALSRDRSDCLGEHLYVMVNAKFEEESKEPVADEEEEDSDDDVEPITEFRFVPSDKSALEAMFTAMCECQALHPDPEDEDSDDYDGEEYDVEAHEQGQGDIPTFYTYEEGLSHLTAEGQATLERLEGMLSQSVSSQYNMAGVRTEDSIRDYEDGMEVDTTPTVAGQFEDADVDH
Specifications
NP_001284 | |
chloride channel, nucleotide-sensitive, 1A | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLNS1A | |
GST |
1207 | |
Wheat germ expression system | |
25 μg | |
CLCI, CLNS1B, ICln | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |