Learn More
Abnova™ CLNS1A (Human) Recombinant Protein
Human CLNS1A full-length ORF ( NP_001284, 1 a.a. - 237 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number NP_001284
Gene ID (Entrez) 1207
Name chloride channel, nucleotide-sensitive, 1A
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16163311
View Documents Promotion Details
Abnova™
H00001207-P01.10UG
10 μg
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
16173311
View Documents Promotion Details
Abnova™
H00001207-P01.25UG
25 μg
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
Description
  • Sequence: MSFLKSFPPPGPAEGLLRQQPDTEAVLNGKGLGTGTLYIAESRLSWLDGSGLGFSLEYPTISLHALSRDRSDCLGEHLYVMVNAKFEEESKEPVADEEEEDSDDDVEPITEFRFVPSDKSALEAMFTAMCECQALHPDPEDEDSDDYDGEEYDVEAHEQGQGDIPTFYTYEEGLSHLTAEGQATLERLEGMLSQSVSSQYNMAGVRTEDSIRDYEDGMEVDTTPTVAGQFEDADVDH
Specifications
Specifications
NP_001284
chloride channel, nucleotide-sensitive, 1A
125% SDS-PAGE Stained with Coomassie Blue
CLCI, CLNS1B, ICln
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
1207
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLNS1A