missing translation for 'onlineSavingsMsg'
Learn More

Clenisohexerol hydrochloride, Dr. Ehrenstorfer

Product Code. 30313801
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313801 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313801 Supplier Dr. Ehrenstorfer Supplier No. DREC11668590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clenisohexerol Hydrochloride
CAS 37158-48-8
Analyte or Component Names Clenisohexerol Hydrochloride
Shipping Condition +20°C
Molecular Formula C14 H22 Cl2 N2 O . Cl H
Certifications/Compliance ISO/IEC 17025
Synonym Clenisohexerol hydrochloride, Benzenemethanol, 4-amino-3,5-dichloro-alpha-[[(1,1-dimethylbutyl)amino]methyl]-, hydrochloride (1:1), Benzenemethanol, 4-amino-3,5-dichloro-alpha-[[(1,1-dimethylbutyl)amino]methyl]-, monohydrochloride (9CI)
InChI Formula InChI=1S/C14H22Cl2N2O.ClH/c1-4-5-14(2,3)18-8-12(19)9-6-10(15)13(17)11(16)7-9;/h6-7,12,18-19H,4-5,8,17H2,1-3H3;1H
SMILES Cl.CCCC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)-2-(2-methylpentan-2-ylamino)ethanol;hydrochloride
Molecular Weight (g/mol) 341.7
Quantity 10 mg
Formula Weight 340.0876 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.