Learn More
(±)-Clenbuterol 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30313297
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313297 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30313297 Supplier Dr. Ehrenstorfer Supplier No. DREA11668560AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (±)-Clenbuterol
CAS 37148-27-9
Analyte or Component Names (±)-Clenbuterol
Shipping Condition Room Temperature
Molecular Formula C12 H18 Cl2 N2 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]-, 4-Amino-3,5-dichloro-α-[[(1,1-dimethylethyl)amino]methyl]benzenemethanol, (RS)-Clenbuterol, (±)-Clenbuterol, 1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol, Clenbuterol, Monores, P 5369, Planipart, dl-Clenbuterol
InChI Formula InChI=1S/C12H18Cl2N2O/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7/h4-5,10,16-17H,6,15H2,1-3H3
SMILES CC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 277.19
Quantity 1 mL
Formula Weight 276.0796 g/mol