missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC3B/Tetranectin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
234.00€ - 307.65€
Specifications
| Antigen | CLEC3B/Tetranectin |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676992
|
Novus Biologicals
NBP2-98344-100ul |
100 μg |
326.00€ 307.65€ / 100µL Save 18.35€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608034
|
Novus Biologicals
NBP2-98344-50ul |
50 μg |
234.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLEC3B/Tetranectin Polyclonal antibody specifically detects CLEC3B/Tetranectin in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| CLEC3B/Tetranectin | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 7123 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, tetranectin (plasminogen binding protein), TNAtetranectin (plasminogen-binding protein), TNtetranectin | |
| Produced in rabbits immunized with purified, recombinant Mouse CLEC3B/Tetranectin (Uniprot#: P43025; Met1-Val202) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title