Learn More
CLEC-2 (CLEC1B) Mouse anti-Human, Brilliant Violet 786, Clone: 219133, BD OptiBuild™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 748143
Product Code. 17219784
  • ONLINE EXCLUSIVE PRICE
    540.00€
    539.91€ / 50µg
    Save 0.09€
    0.02% Off
Description
CLEC-2 (CLEC1B) Monoclonal antibody specifically detects CLEC-2 (CLEC1B) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLEC-2 (CLEC1B)
Monoclonal
0.2 mg/mL
Mouse
RUO
CLEC-2 (CLEC1B)
Refrigerate (2 to 8°C)
Flow Cytometry
219133
Brilliant Violet 786
50 μg
Primary
Human
IgG2a κ