Learn More
Fisherbrand™ Soda Lime Glass Bottle with 28mm Fitted Closure, Clear
Product Code. 12820884 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
100 mL
150 mL
300 mL
1000 mL
Unit Size:
Pack of 10
Pack of 20
Pack of 40
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12820884 150 mL Pack of 20
12890874 30 mL Pack of 40
12800884 60 mL Pack of 40
12810884 100 mL Pack of 20
12840884 300 mL Pack of 20
12860884 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12820884
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use the Fisherbrand™ Soda Lime Glass Bottle with 28mm Fitted Closure, Clear in the laboratory to store and manipulate laboratory media for experimentation.
  • Tamper evident vistop closure with polycone liner
  • ISO certified
  • Clear soda lime glass
Specifications
Capacity (Metric) 150 mL
Color Clear
Diameter (Metric) Inner 51.6 mm
Fill Volume Range 158mL
Material Soda Lime Glass
Neck ID (Metric) 28mm
No. per Pack 20Pack
Quantity 20/Pk
Screw Material Polypropylene
Closure Color White
Liner Material EPE
Closure Included Yes
Shape Round
Height (Metric) 123 mm
Product Type Screw Neck Bottle
Closure Liner Polypropylene/SOR
Closure Material Polypropylene
Diameter (Metric) 51.6 mm