Learn More
PURUS™ Cleantek CPE Overshoes
Product Code. 15608643 Shop All Purus Products
Change view
Click to view available options
Length (Metric):
355.6 mm
406.4 mm
Unit Size:
Pack of 2000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15608643 355.6 mm Pack of 2000
15618643 406.4 mm Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15608643 Supplier PURUS™ Supplier No. 48020814
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Disposable shoe covers provide barrier protection against light spills and dust
  • 100 pieces per bag, 20 per case
  • Made from cast polyethelyne
  • helps reduce particulates from entering a cleanroom environment
  • provides protection for shoes against light splash and dust
Specifications
Color Blue
Length (English) 14 in.
Length (Metric) 355.6 mm
Disposable Single-use
Packaging Quantity 2,000 Pc./Case