missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLDND1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 463.00€
Specifications
| Antigen | CLDND1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656100
|
Novus Biologicals
NBP2-92420-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688200
|
Novus Biologicals
NBP2-92420-0.1ml |
0.1 mL |
463.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLDND1 Polyclonal antibody specifically detects CLDND1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| CLDND1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 56650 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| C3orf4, chromosome 3 open reading frame 4, claudin domain containing 1, claudin domain containing 1 protein, claudin domain-containing protein 1, GENX-3745, Membrane protein GENX-3745, MGC111162, MGC3316, MGC9861 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-138 of human CLDND1 (Q9NY35). GTDFWYEYRSPVQENSSDLNKSIWDEFISDEADEKTYNDALFRYNGTVGLWRRCITIPKNMHWYSPPERTESFDVVTKCVSFTLTEQFMEKFVDPGNHNSGIDLLRTYLWR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title