Learn More
CLC-5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA577369
Product Code. 15943875
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CLC-5
Polyclonal
Unconjugated
Clcn5
CLCN5
Rabbit
Antigen affinity chromatography
RUO
12728, 25749
-20°C
Lyophilized
Immunohistochemistry, Western Blot
0.8 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.4
P51796, Q9WVD4
Clcn5
Peptide CDYENHFNTSKGGEL, corresponding to amino acid residues 401-415 of rat CLC5
50 μL
Primary
Mouse, Rat
Antibody