missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00€ - 322.00€
Specifications
| Antigen | Claudin-6 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616067
|
Novus Biologicals
NBP3-12787-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620687
|
Novus Biologicals
NBP3-12787-50ul |
50 μg |
195.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin-6 Polyclonal antibody specifically detects Claudin-6 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Claudin-6 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 9074 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| claudin 6, claudin-6, skullin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse Claudin-6 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title