missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-18.2 Antibody (CLDN18.2/8559R), Novus Biologicals™
Rabbit Monoclonal Antibody
291.00€ - 530.00€
Specifications
| Antigen | Claudin-18.2 |
|---|---|
| Clone | CLDN18.2/8559R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641795
|
Novus Biologicals
NBP3-23718-20ug |
20 μg |
291.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616854
|
Novus Biologicals
NBP3-23718-100ug |
100 μg |
530.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin-18.2 Monoclonal antibody specifically detects Claudin-18.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Claudin-18.2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 51208 | |
| IgG κ | |
| Protein A or G purified |
| CLDN18.2/8559R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| claudin-18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J | |
| Recombinant fragment (around aa1-100) of human Claudin-18.2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title