missing translation for 'onlineSavingsMsg'
Learn More

Claudin-18.2 Antibody (CLDN18.2/7300R), Novus Biologicals™

Rabbit Monoclonal Antibody

291.00€ - 519.00€

Specifications

Antigen Claudin-18.2
Clone CLDN18.2/7300R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18600115
View Documents
Novus Biologicals
NBP3-23700-100ug
100 μg
519.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18656574
View Documents
Novus Biologicals
NBP3-23700-20ug
20 μg
291.00€
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Claudin-18.2 Monoclonal antibody specifically detects Claudin-18.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Claudin-18.2
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Signal Transduction
10 mM PBS with 0.05% BSA
51208
IgG κ
Protein A or G purified
CLDN18.2/7300R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
claudin-18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J
Recombinant fragment (around aa1-100) of human Claudin-18.2 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.