missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 15 Monoclonal Antibody (540315)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560879
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Plays a major role in tight junction specific obliteration of the intercellular space, through calcium independent cell adhesion activity.
Specifications
| Claudin 15 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P56746 | |
| CLDN15 | |
| NS0 mouse myeloma cell line transfected with human Claudin-15. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 540315 | |
| Unconjugated | |
| CLDN15 | |
| 2210009B08Rik; BB107105; claudin 15; claudin-15; CLDN15 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 24146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction