missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claudin 12 Monoclonal Antibody (393314)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561412

Product Code. 30535740

  • 469.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Claudin 12
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
P56749
CLDN12
HEK293 human embryonic kidney cell line transfected with human Claudin-12, Met1-Thr244.
100 μg
Primary
Human
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry
393314
Unconjugated
CLDN12
claudin 12; claudin-12; Cldn12; RGD1561053
Mouse
Protein A/G
RUO
9069
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.