missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin Heavy Chain 1/CHC17 Antibody (CLTC/1431), Novus Biologicals™
Mouse Monoclonal Antibody
264.00€ - 518.00€
Specifications
| Antigen | Clathrin Heavy Chain 1/CHC17 |
|---|---|
| Clone | CLTC/1431 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18189819
|
Novus Biologicals
NBP2-53255-20UG |
20 μg |
264.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18297160
|
Novus Biologicals
NBP2-53255-100UG |
100 μg |
518.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human samples. It is validated for ELISA.Specifications
| Clathrin Heavy Chain 1/CHC17 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 1213 | |
| Purified recombinant N-terminal fragment of human Clathrin Heavy Chain (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| CLTC/1431 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034 | |
| CLTC | |
| IgG1 κ | |
| Protein A or G purified | |
| 192 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title