missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claspin Monoclonal Antibody (485508)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561295

Product Code. 30535004

  • 459.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Claspin
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9HAW4
CLSPN
E. coli-derived recombinant human Claspin, Phe1138-Ser1339.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
485508
Unconjugated
CLSPN
B130025E01; C85083; claspin; claspin homolog; CLSPN; E130314M08Rik; hClaspin; MGC131612; MGC131613; MGC131615
Mouse
Protein A/G
RUO
63967
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.