missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Citalopram, 5-HT Reuptake Inhibitor, >98% Purity, Solid, 25 mg
Description
- Molecular Weight: 405.3 Da
- Purity: >98%
- Description: Selective serotonin reuptake inhibitor, acts as an antidepressant in vivo
- Form: Solid
- Chemical Name: 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile hydrobromide
- Storage: Store at ambient
- Shipping: Ambient - Can ship with ice
- Citation Count: 5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | 5-HT (Serotonin) |
| Formulation | 100% 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile hydrobromide |
| Quantity | 25 mg |
| For Use With (Application) | Antidepressant research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?