missing translation for 'onlineSavingsMsg'
Learn More

(±)-cis-Nerolidol 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30327967
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327967 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327967 Supplier Dr. Ehrenstorfer Supplier No. DREA15503030ME100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-Nerolidol
CAS 3790-78-1
Analyte or Component Names cis-Nerolidol
Shipping Condition Room Temperature
Molecular Formula C15 H26 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,6,10-Dodecatrien-3-ol, 3,7,11-trimethyl-, (6Z)-, 1,6,10-Dodecatrien-3-ol, 3,7,11-trimethyl-, (Z)- (8CI), (6Z)-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, (Z)-Nerolidol, (±)-cis-Nerolidol, cis-Nerolidol
InChI Formula InChI=1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11-
SMILES CC(=CCC\C(=C/CCC(C)(O)C=C)\C)C
IUPAC Name (6Z)-3,7,11-trimethyldodeca-1,6,10-trien-3-ol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 222.37
Quantity 1 mL
Formula Weight 222.1984 g/mol
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.