missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CINAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | CINAP |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:300-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689521
|
Novus Biologicals
NBP2-98094-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604335
|
Novus Biologicals
NBP2-98094-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CINAP Polyclonal antibody specifically detects CINAP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CINAP | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Neuroscience | |
| PBS (pH 7.0) | |
| 64061 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:300-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CASK-interacting nucleosome assembly protein, CDA1CTCL-associated antigen se20-4, Cell division autoantigen 1, CINAPSE20-4, CTCL, CTCL tumor antigen se20-4, Cutaneous T-cell lymphoma-associated antigen se20-4, cutaneous T-cell lymphoma-associated tumor antigen se20-4, DENTTTSPY-like protein 2, differentially expressed nucleolar TGF-beta1 target, Differentially-expressed nucleolar TGF-beta1 target protein, HRIHFB2216, NP79, Nuclear protein of 79 kDa, SE204, testis-specific protein Y encoded-like 2, testis-specific Y-encoded-like protein 2, TSPX, TSPY-like 2 | |
| Produced in rabbits immunized with E. coli-derived Human CINAP fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title