missing translation for 'onlineSavingsMsg'
Learn More

CIITA Monoclonal Antibody (OTI7B12), TrueMAB™, OriGene

Product Code. 17556544
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17556544 30 μL 30µL
17536544 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17556544 Supplier Origene Technologies Supplier No. TA812200S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CIITA Monoclonal antibody specifically detects CIITA in Human samples. It is validated for Western Blot

Specifications

Antigen CIITA
Applications Western Blot
Classification Monoclonal
Clone OTI7B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ACTR3
Gene Accession No. P61158
Gene Alias C2ta; CIITA; CIITAIV; class II major histocompatibility complex transactivator; class II transactivator; class II, major histocompatibility complex, transactivator; EG669998; Gm9475; MH NLRA; MHC class II transactivator; MHC class II transactivator type I; MHC class II transactivator type III; MHC class II transactivator type IV; MHC2TA; NLR family, acid domain containing; NLRA; nucleotide-binding oligomerization domain, leucine rich repeat and acid domain containing
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-418 of human ACTR3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10096
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.