missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Antibody (CHGA/765), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47847F
This item is not returnable.
View return policy
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Chromogranin A | |
| Monoclonal | |
| FITC | |
| betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I | |
| Recombinant human Chromogranin A protein (Uniprot: P10645) | |
| 0.1 mL | |
| Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers | |
| 1113 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CHGA/765 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction