missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Ab-1 Mouse Monoclonal Antibody, Epredia™

Product Code. 12691007 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
Unit Size:
100µL
1mL
200µL
500µL
7mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate Form Concentration unitSize
12691007 500 μL Unconjugated Concentrated 200μg/mL 500µL
12651007 100 μL Biotin Concentrated 200μg/mL 100µL
12661007 500 μL Biotin Concentrated 200μg/mL 500µL
12681007 100 μL Unconjugated Concentrated 200μg/mL 100µL
12601017 100 μL Unconjugated Concentrated 1mg/mL 100µL
12611017 200 μL Unconjugated Concentrated 1mg/mL 200µL
12671007 1 mL Unconjugated Concentrated 200μg/mL 1mL
12641017 7 mL Unconjugated Prediluted Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12691007 Supplier Epredia Supplier No. MS324P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.

Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: LK2H10

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: PC-12 cells, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromogranin A Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LK2H10
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human pheochromocytoma
Quantity 500 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Non-human Primate, Pig, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.