Learn More
Macherey-Nagel™ Chrombond™ RS 80 C18 ec Polypropylene Flash Column
Product Code. 12738332
Click to view available options
:
I.D.: 30.8mm
Product Code. 12738332
Brand: Macherey-Nagel™ 732814
Only null left
Specifications
Length (Metric) 240 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Directly Used in The Teledyne Isco Companion, Rf