Learn More
Macherey-Nagel™ Chromafix™ SiOH Cartridge
Product Code. 10023031
Change view
Click to view available options
Description:
Adsorbent Weight: 230 mg
Adsorbent Weight: 420 mg
Adsorbent Weight: 880 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10023031 Adsorbent Weight: 880 mg Pack of 50
10063191 Adsorbent Weight: 230 mg Pack of 50
10411862 Adsorbent Weight: 420 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10023031 Supplier Macherey-Nagel™ Supplier No. 731830
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
Estimated Shipment: 10-09-2026
Specifications
Base Material Weakly Acidic Silica
Description Adsorbent Weight: 880 mg
For Use With (Application) Aflatoxins, Chloramphenicol, Pesticides, Steroids, Vitamins
Particle Size 45 μm
Sorbent SiOH
Type Chromafix SiOH Cartridge
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g