Learn More
Macherey-Nagel™ Chromafix™ PS-Ba2+ cartridges
Specifications
Specifications
| Base Material | Strong PS/DVB Cation Exchanger, Ba2+ form |
| Bed Weight | 550 mg |
| For Use With (Application) | Removal of Sulfate Ions From Water |
| Particle Size | 100 μm |
| Type | Chromafix PS-Ba2+ Cartridge |
| Pore Size | 100 Å |
| Quantity | 50 Pack |