Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Frit
Click to view available options
Volume (Metric):
1 mL
3 mL
6 mL
15 mL
30 mL
45 mL
70 mL
150 mL
Unit Size:
Pack of 250
Description
Accessory for SPE (solid-phase extraction)
For individual packing of SPE columns with Chromabond™ adsorbents
Specifications
Specifications
| For Use With (Equipment) | Polypropylene Columns |
| Type | Frit |
| Volume (Metric) | 70 mL |
| Material | Polyethylene |
| Quantity | 250 Pack |