Learn More
Macherey-Nagel™ Chromabond™ BT 80 SiOH Column
Product Code. 15144202
Change view
Click to view available options
Length (Metric):
240 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15144202 240 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15144202 Supplier Macherey-Nagel™ Supplier No. 732964
In Stock
Specifications
Length (Metric) 240 mm
Type Flash Column
Diameter (Metric) Outer 30.8 mm
For Use With (Equipment) Biotage Isolera Systems