missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRM3 Monoclonal Antibody (580011)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561658

Product Code. 30534151

  • 469.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 3 controls smooth muscle contraction and its stimulation causes secretion of glandular tissue.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CHRM3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
P20309
CHRM3
NS0 mouse myeloma cell line transfected with human CHRM3.
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin)
580011
Unconjugated
CHRM3
acetylcholine receptor; acetylcholine receptor, muscarinic 3; AChR M3; cholinergic receptor; cholinergic receptor muscarinic 3; cholinergic receptor, muscarinic 3; cholinergic receptor, muscarinic 3, cardiac; CHRM3; Chrm-3; EGBRS; HM3; M3; m3 muscarinic receptor; M3R; Mm3 mAChR; muscarinic acetylcholine receptor 3; muscarinic acetylcholine receptor M3; PBS
Mouse
Protein A/G
RUO
1131
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.