missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRFAM7A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | CHRFAM7A |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230085
|
Novus Biologicals
NBP3-38474-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228862
|
Novus Biologicals
NBP3-38474-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHRFAM7A Polyclonal antibody specifically detects CHRFAM7A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| CHRFAM7A | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CHRNA7, CHRNA7 (cholinergic receptor, nicotinic, alpha 7, exons 5-10) and FAM7A (familywith sequence similarity 7A, exons A-E) fusion, CHRNA7 (cholinergic receptor, nicotinic, alpha polypeptide 7, exons 5-10) andFAM7A (family with sequence similarity 7A, exons A-E) fusion, CHRNA7-DR1alpha 7 neuronal nicotinic acetylcholine receptor-FAM7A hybrid, CHRNA7-FAM7A fusion protein, D-10alpha-7 nicotinic cholinergic receptor subunit, MGC120482, MGC120483 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human CHRFAM7A (NP_647536.1).,, Sequence:, MQKYCIYQHFQFQLLIQHLWIAANCDIADERFDATFHTNVLVNSSGHCQYLPPGIFKSSCYIDVRWFPFDVQHCKLKFGSWSYGGWSLDLQMQEADISGYIPNGEWDLVGIPGKRSERFYECCKEPYPDVTFTVTMRRRTLYYGLNLLIP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 89832 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title