missing translation for 'onlineSavingsMsg'
Learn More

CHRAC1 Antibody, Novus Biologicals™

Product Code. 30232191 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30232191 20 μL 20µL
30227294 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30232191 Supplier Novus Biologicals Supplier No. NBP33795020ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHRAC1 Polyclonal antibody specifically detects CHRAC1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRAC1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias CHARC1, CHRAC-1, CHRAC-15, CHRAC15CHARC15, chromatin accessibility complex 1, Chromatin accessibility complex 15 kDa protein, DNA polymerase epsilon subunit p15, histone-fold protein CHRAC15, HuCHRAC15, YCL1chromatin accessibility complex protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-131 of human CHRAC1 (NP_059140.1).,, Sequence:, MADVVVGKDKGGEQRLISLPLSRIRVIMKSSPEVSSINQEALVLTAKATELFVQCLATYSYRHGSGKEKKVLTYSDLANTAQQSETFQFLADILPKKILASKYLKMLKEEKREEDEENDNDNESDHDEADS
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 54108
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.