missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHOP; GADD153 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81462-1-RR-20UL
This item is not returnable.
View return policy
Description
The CHOP; GADD153 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CHOP; GADD153. This antibody recognizes human antigen. The CHOP; GADD153 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| CHOP; GADD153 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| CEBPZ; CHOP; CHOP10; GADD153; MGC4154 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 30 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1G11 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 1649 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction