missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorine Standard: Cl @ 200 μg/g (0.0200 wt%) in 75 cSt Mineral Oil, VHG™
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
Chlorine Standard: Cl @ 200 μg/g (0.0200 wt%) in 75 cSt Mineral Oil
Specifications
Specifications
| Chemical Name or Material | Chlorine |
| CAS | 7782-50-5 |
| Analyte or Component Names | Chlorine |
| Shipping Condition | Room Temperature |
| Molecular Formula | Cl2 |
| Concentration | 200 μg/g |
| Synonym | Cl |
| InChI Formula | InChI=1S/Cl2/c1-2 |
| SMILES | ClCl |
| IUPAC Name | molecular chlorine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction