missing translation for 'onlineSavingsMsg'
Learn More

Chlorine - Cl @ 5000 μg/g in 75 cSt Mineral Oil

Product Code. 30325553
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 30325553

Brand: VHG Labs VHGCLOIL5000500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorine
CAS 7782-50-5
Analyte or Component Names Chlorine
Shipping Condition Room Temperature
Molecular Formula Cl2
Concentration 5000 μg/g
Synonym Cl
InChI Formula InChI=1 S/Cl2/c1-2
Solution Type Single Solution
SMILES ClCl
Solvent or Matrix 75 cSt Mineral Oil
Industry Type Industrial QC
Molecular Weight (g/mol) 70.91
Quantity 500 mL
Formula Weight 69.9377
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.