missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhexidine Dihydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Chlorhexidine dihydrochloride |
| CAS | 3697-42-5 |
| Analyte or Component Names | Chlorhexidine dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H30 Cl2 N10 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Chlorhexidine Dihydrochloride, N1,N14-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide hydrochloride (1:2), N,N''-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide dihydrochloride, 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] dihydrochloride, AY 5312, Arlacide H, Chlorhexidine dihydrochloride, Chlorhexidine hydrochloride, Chlorhexidinium dichloride, Chlorohexidine dihydrochloride, Lisium, Sebidin |
| InChI Formula | InChI=1S/C22H30Cl2N10.2ClH/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;;/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H |
| Solution Type | Neat |
| SMILES | Cl.Cl.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?