missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase 3-like 2 Antibody (YKL39/6370), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45073
This item is not returnable.
View return policy
Description
Chitinase 3-like 2 Monoclonal antibody specifically detects Chitinase 3-like 2 in Human samples. It is validated for ELISA,Functional Assay
Specifications
| Chitinase 3-like 2 | |
| Monoclonal | |
| 1 mg/mL | |
| 10 mM PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG2 κ |
| ELISA, Functional Assay | |
| YKL39/6370 | |
| Unconjugated | |
| chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL-39YKL39 | |
| Recombinant human fragment corresponding to Chitinase 3-like 2 protein (Uniprot: Q15782). Exact sequence is proprietary. | |
| 100 μg | |
| Stem Cells | |
| 1117 | |
| Store at -20 to -84°C. Avoid freeze-thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction