missing translation for 'onlineSavingsMsg'
Learn More

Chikungunya Virus E2 Antibody Pair [HRP], Novus Biologicals™

Product Code. 31849945 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31849945 5 Plates -
31844987 15 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31849945 Supplier Novus Biologicals Supplier No. NBP3491685Plates
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Chikungunya Virus E2
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus E2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Gene Alias Chikungunya, E2 envelope glycoprotein, P130, Spike glycoprotein E2
Host Species Mouse
Quantity 5 Plates
Regulatory Status RUO
Target Species Virus
Content And Storage Storage of components varies. See protocol for specific instructions.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.