missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chikungunya Virus E1 Monoclonal Antibody (988110)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562051
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Chikungunya fever is a pandemic disease caused by the mosquito-borne Chikungunya virus (CHIKV). The virus E1 glycoprotein mediation of viral membrane fusion during CHIKV infection is a crucial step in the release of viral genome into the host cytoplasm for replication.
Specifications
| Chikungunya Virus E1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA | |
| 988110 | |
| Unconjugated | |
| Chikungunya Virus | |
| Human embryonic kidney cell line HEK293-derived transfected with viral Chikungunya Virus E1, Tyr810-Gly1224. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction