missing translation for 'onlineSavingsMsg'
Learn More

CHID1 Antibody (CHID1/6368), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18552748 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18552748 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18552748 Supplier Novus Biologicals Supplier No. NBP324214MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CHID1 Monoclonal antibody specifically detects CHID1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHID1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CHID1/6368
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias chitinase domain containing 1, chitinase domain-containing protein 1, FLJ42707, MGC3234, SI-CLP, stabilin-1 interacting chitinase-like protein, Stabilin-1-interacting chitinase-like protein
Host Species Mouse
Immunogen Recombinant full-length human CHID1 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 66005
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.