missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHIA Monoclonal Antibody (OTI3D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811213
This item is not returnable.
View return policy
Description
Degrades chitin and chitotriose. May participate in the defense against nematodes, fungi and other pathogens. Plays a role in T helper cell type 2 (Th2) immune response. Contributes to the response to IL-13 and inflammation in response to IL-13. Stimulates chemokine production by pulmonary epithelial cells. Protects lung epithelial cells against apoptosis and promotes phosphorylation of AKT1. Its function in the inflammatory response and in protecting cells against apoptosis is inhibited by allosamidin, suggesting that the function of this protein depends on carbohydrate binding.Specifications
| CHIA | |
| Monoclonal | |
| Unconjugated | |
| CHIA | |
| AMCASE, CHIT2, TSA1902 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3D8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BZP6 | |
| CHIA | |
| Full length human recombit protein of human CHIA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction