missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHIA Monoclonal Antibody (OTI3D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811213S
This item is not returnable.
View return policy
Description
CHIA Monoclonal antibody specifically detects CHIA in Human, Mouse samples. It is validated for Western BlotSpecifications
| CHIA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99LE6, Q9UG63 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10061, 27407 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI3D8 | |
| Unconjugated | |
| Abcf2 | |
| 2200003E03Rik; acidic chitinase; acidic mammalian chitinase; AMCASE; Chia; Chia1; CHIT2; chitinase, acidic; chitinase, acidic 1; eosinophil chemotactic cytokine; lung-specific protein TSA1902; small acid mammalian chitinase; TSA1902; YNL | |
| Human recombinant protein fragment corresponding to amino acids 1-235 of human ABCF2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction