missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChemR23/CMKLR1 Antibody (BZ194) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
409.00€ - 695.00€
Specifications
| Antigen | ChemR23/CMKLR1 |
|---|---|
| Clone | BZ194 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18503570
|
Novus Biologicals
NBP1-43233-0.5mg |
0.5 mg |
695.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18564203
|
Novus Biologicals
NBP1-43233-0.1mg |
0.1 mg |
481.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18518170
|
Novus Biologicals
NBP1-43233-0.05mg |
0.05 mg |
409.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ChemR23/CMKLR1 Monoclonal antibody specifically detects ChemR23/CMKLR1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ChemR23/CMKLR1 | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| GPCR, Immunology | |
| PBS | |
| CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, DEZ, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
| Partial synthetic peptide made to an N-terminal portion of the mouse ChemR23/CMKLR1 protein (amino acids 8-32) [UniProt P97468] | |
| Primary | |
| Store at 4°C. Do not freeze. |
| BZ194 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P97468 | |
| 1240 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title