missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92563-0.02ml
This item is not returnable.
View return policy
Description
CHD5 Polyclonal antibody specifically detects CHD5 in Human samples. It is validated for Western Blot
Specifications
| CHD5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATP-dependent helicase CHD5, CHD-5, chromodomain helicase DNA binding protein 5, chromodomain-helicase-DNA-binding protein 5, DKFZp434N231, EC 3.6.1, EC 3.6.4.12, KIAA0444 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1530-1700 of human CHD5 (NP_056372.1). IPEGPEGKKSGEVISSDPNTPVPASPAHLLPAPLGLPDKMEAQLGYMDEKDPGAQKPRQPLEVQALPAALDRVESEDKHESPASKERAREERPEETEKAPPSPEQLPREEVLPEKEKILDKLELSLIHSRGDSSELRPDDTKAEEKEPIETQQNGDKEEDDEGKKEDKKGK | |
| 0.02 mL | |
| Chromatin Modifiers | |
| 26038 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction