missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | CHD4 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688115
|
Novus Biologicals
NBP2-99098-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649825
|
Novus Biologicals
NBP2-99098-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHD4 Polyclonal antibody specifically detects CHD4 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CHD4 | |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immune System Diseases, Immunology | |
| 0.2 μm filtered solution in PBS | |
| 1108 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CHD4. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title