missing translation for 'onlineSavingsMsg'
Learn More

cGK2/PRKG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00€ - 550.00€

Specifications

Antigen cGK2/PRKG2
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug-4ug antibody for 400ug-600ug extracts of whole cells
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30231188
View Documents
Novus Biologicals
NBP3-35308-100ul
100 μL
550.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30231190
View Documents
Novus Biologicals
NBP3-35308-20ul
20 μL
190.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

cGK2/PRKG2 Polyclonal antibody specifically detects cGK2/PRKG2 in Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications

Specifications

cGK2/PRKG2
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Cell Cycle and Replication, Phospho Specific
PBS (pH 7.3), 50% glycerol
5593
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug-4ug antibody for 400ug-600ug extracts of whole cells
Polyclonal
Purified
RUO
Mouse, Rat
cGK 2, cGKIIcGK2, cGMP-dependent protein kinase 2, cGMP-dependent protein kinase II, EC 2.7.11, PRKGR2EC 2.7.11.12, protein kinase, cGMP-dependent, type II
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human cGK2/PRKG2 (NP_006250.1).,, Sequence:, MGNGSVKPKHSKHPDGHSGNLTTDALRNKVTELERELRRKDAEIQEREYHLKELREQLSKQTVAIAELTEELQNKCIQLNKLQDVVHMQGGSPLQASPDKVPLEVHRKTSGLVSLHSRRGAKAGVSAEPTTRTYDLNKPPEFSFEKARVRKDSSEKKLIT
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.