missing translation for 'onlineSavingsMsg'
Learn More

CGI-09 Antibody, Novus Biologicals™

Product Code. 18293883 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18293883 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18293883 Supplier Novus Biologicals Supplier No. NBP150039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CGI-09 Polyclonal specifically detects CGI-09 in Human samples. It is validated for Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CGI-09
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 mcg/mg
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias CGI-09, GCD10, Gcd10p, KIAA1153, MGC5029, TRM6, tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6, tRNA methyltransferase 6 homolog (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM6, tRNA(m1A58)MTase subunit TRM6
Gene Symbols TRMT6
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 325 and 375 of human tRNA Methyltransferase 6 using the numbering given in entry NP_057023.2 (GeneID 51605).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51605
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.