Learn More
CFP Mouse, anti-Human, CoraLite Plus 488, Clone: 1D5A9, Proteintech
Product Code. 18098175 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18098175 20 μL 20µL
18035295 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18098175 Supplier Proteintech Supplier No. CL4886678820UL
 Restricted Item
Estimated Shipment: 21-10-2026
Mouse Monoclonal Antibody
The CFP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CFP. This antibody recognizes human, pig, rat antigen. The CFP antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Antigen CFP
Applications Western Blot
Clone 1D5A9
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols BFD, CFP, Complement factor P, complement factor properdin, PFC, PFD, PROPERDIN
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 5199
Target Species Human, Pig, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 155 kDa
Form Liquid
Isotype IgG1