missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CFHR1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148821
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a secreted protein belonging to the complement factor H protein family. It binds to Pseudomonas aeruginosa elongation factor Tuf together with plasminogen, which is proteolytically activated. It is proposed that Tuf acts as a virulence factor by acquiring host proteins to the pathogen surface, controlling complement, and facilitating tissue invasion. Mutations in this gene are associated with an increased risk of atypical hemolytic-uremic syndrome.
Specifications
| CFHR1 | |
| Polyclonal | |
| Unconjugated | |
| CFHR1 | |
| AI194696; CFHL; Cfhl1; CFHL1P; CFHR1; CFHR1P; CFHRB; complement component factor h-like 1; complement factor H related 1; complement factor H-related 1; complement factor H-related 1 pseudogene; complement factor H-related protein; complement factor H-related protein 1; FHR1; FHR-1; H factor (complement)-like 1; H factor (complement)-like 2; h factor-like protein 1; H36; H36-1; H36-2; H-factor-like 1; HFL1; HFL2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 3078 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q03591 | |
| CFHR1 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 1/CFHR1, Glu19-Ala328. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction